Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122683.1 | NP_001116155.1 | 7 | 1132 | ||
NM_175177.4 | NP_780386.3 | 7 | 1113 | |||
GenBank mRNA | AK009575.1 | - | 7 | 1090 | ||
AK076718.1 | - | 7 | 1086 | |||
AK137955.1 | - | 7 | 1073 | |||
AK145711.1 | - | 7 | 1133 | |||
AK146321.1 | - | 7 | 1137 | |||
AK146962.1 | - | 8 | 1182 | |||
BC027063.1 | - | 3 | 588 | |||
BC043683.1 | - | 7 | 1116 | 5133856 | ||
BC096457.1 | - | 7 | 1061 | 4975979 | ||
EF095210.1 | - | 1 | 202 |