Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dus4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028002.2 | NP_082278.1 | 6 | 665 | ||
GenBank mRNA | AK010138.1 | - | 6 | 666 | ||
BC109351.2 | - | 6 | 600 | 40056894 |