Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itfg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028007.3 | NP_082283.2 | 11 | 1369 | ||
GenBank mRNA | AK009867.1 | - | 11 | 1193 | ||
AK040124.1 | - | 1 | 2234 | |||
AK132242.1 | - | 11 | 1315 | |||
AK141948.1 | - | 11 | 1341 | |||
AK210719.1 | - | 1 | 67 | |||
BC003977.1 | - | 11 | 1329 | 3489518 |