Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itfg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028007.3 | NP_082283.2 | 5, 6 | 761 | ||
GenBank mRNA | AK009867.1 | - | 5, 6 | 585 | ||
AK132242.1 | - | 5, 6 | 706 | |||
AK141948.1 | - | 5, 6 | 733 | |||
AK178165.1 | - | 1, 2 | 12 | |||
BC003977.1 | - | 5, 6 | 721 | 3489518 |