Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024495.5 | NP_078771.1 | 4 | 515 | ||
GenBank mRNA | AF231123.1 | - | 4 | 465 | ||
AK010166.1 | - | 4 | 469 | |||
AK150871.1 | - | 4 | 467 | |||
AK150897.1 | - | 4 | 467 | |||
AK151519.1 | - | 4 | 453 | |||
AK151978.1 | - | 4 | 467 | |||
AK152124.1 | - | 4 | 464 | |||
AK153080.1 | - | 4 | 494 | |||
AK153258.1 | - | 4 | 455 | |||
AK153353.1 | - | 4 | 494 | |||
AK162621.1 | - | 4 | 515 | |||
BC064050.1 | - | 4 | 466 | 949505 |