Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310067B10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028014.3 | NP_082290.2 | 27 | 3676 | ||
GenBank mRNA | AK047622.1 | - | 13 | 1810 | ||
BC007157.1 | - | 14 | 1922 | |||
BC030865.1 | - | 12 | 1443 | |||
BC053088.1 | - | 27 | 3591 | 5709104 |