Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028019.3 | NP_082295.1 | 1 | 101 | ||
GenBank mRNA | AK002781.1 | - | 1 | 79 | ||
AK010429.1 | - | 1 | 103 | |||
AK150921.1 | - | 1 | 102 | |||
AK152096.1 | - | 1 | 94 | |||
BC096385.1 | - | 1 | 95 | 30744704 | ||
BC132307.1 | - | 1 | 97 | 40130584 | ||
BC138303.1 | - | 1 | 97 | 8861322 | ||
BC145236.1 | - | 1 | 96 |