Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbed5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183088.2 | NP_898911.2 | 4 | 1793 | ||
GenBank mRNA | AK010551.1 | - | 4 | 1794 | ||
BC054117.1 | - | 1 | 302 | |||
BC137610.1 | - | 4 | 1791 | 8860629 |