Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028037.4 | NP_082313.2 | 9, 10 | 1485 | ||
GenBank mRNA | AK050332.1 | - | 9, 10 | 1689 | ||
AK148195.1 | - | 8, 9 | 1294 | |||
BC027825.1 | - | 9, 10 | 1461 | 5343012 | ||
BC029047.1 | - | 5, 6 | 634 |