Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpusd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028040.2 | NP_082316.1 | 7 | 1081 | ||
GenBank mRNA | AK010322.1 | - | 7 | 1098 | ||
AK156574.1 | - | 7 | 1098 | |||
AK215753.1 | - | 1 | 22 | |||
BC061256.1 | - | 7 | 1082 | 30251586 |