Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028041.2 | NP_082317.1 | 11 | 1140 | ||
GenBank mRNA | AF319547.1 | - | 11 | 1500 | ||
AK010484.1 | - | 8 | 720 | |||
AK028274.1 | - | 11 | 1154 | |||
AK031534.1 | - | 11 | 1150 | |||
AK031679.1 | - | 11 | 1158 | |||
AK147715.1 | - | 11 | 1155 | |||
AK152989.1 | - | 11 | 1156 | |||
AK155138.1 | - | 11 | 1152 | |||
AK167603.1 | - | 11 | 1154 | |||
AK168455.1 | - | 11 | 1154 | |||
AK169255.1 | - | 11 | 1153 | |||
AK170967.1 | - | 11 | 1152 | |||
BC021900.1 | - | 2 | 145 | |||
BC043699.1 | - | 11 | 1140 | 5344158 | ||
BC055026.1 | - | 7 | 583 | |||
BC062920.1 | - | 11 | 1143 | |||
BC071251.1 | - | 5 | 437 |