Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028049.2 | NP_082325.2 | 3, 4 | 489 | ||
GenBank mRNA | AK002745.1 | - | 3, 4 | 369 | ||
AK005473.1 | - | 2, 3 | 317 | |||
AK181573.1 | - | 2, 3 | 149 | |||
AK181749.1 | - | 2, 3 | 150 | |||
AK184767.1 | - | 2, 3 | 149 | |||
BC018273.1 | - | 3, 4 | 429 | 4458296 |