Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028063.2 | NP_082339.1 | 5 | 606 | ||
GenBank mRNA | AK005541.1 | - | 5 | 606 | ||
AK043532.1 | - | 5 | 613 | |||
AK044059.1 | - | 5 | 613 | |||
AY349617.1 | - | 5 | 613 | |||
BC051548.1 | - | 5 | 607 | |||
BC061026.1 | - | 5 | 593 | 30305575 |