Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252578.1 | NP_001239507.1 | 18 | 2883 | ||
NM_001252579.1 | NP_001239508.1 | 18 | 2729 | |||
NM_028072.5 | NP_082348.2 | 18 | 2725 | |||
GenBank mRNA | AK008108.1 | - | 9 | 988 | ||
AK028874.1 | - | 18 | 2640 | |||
AK034712.1 | - | 18 | 2710 | |||
AK036685.1 | - | 18 | 2661 | |||
AK049170.1 | - | 13 | 1640 | |||
AK081643.1 | - | 18 | 2683 | |||
AK129316.1 | - | 18 | 2883 | |||
AK133336.1 | - | 2699 | ||||
AK165183.1 | - | 18 | 2637 | |||
AY101177.1 | - | 17 | 2544 | |||
BC027238.1 | - | 7 | 699 | |||
BC062900.1 | - | 18 | 2681 | |||
BC141086.1 | - | 17 | 2478 | 9055698 |