Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252578.1 | NP_001239507.1 | 10 | 1785 | ||
NM_001252579.1 | NP_001239508.1 | 10 | 1631 | |||
NM_028072.5 | NP_082348.2 | 10 | 1627 | |||
GenBank mRNA | AK028874.1 | - | 10 | 1542 | ||
AK034712.1 | - | 10 | 1612 | |||
AK036685.1 | - | 10 | 1563 | |||
AK049170.1 | - | 5 | 542 | |||
AK081643.1 | - | 10 | 1585 | |||
AK129316.1 | - | 10 | 1785 | |||
AK133336.1 | - | 1601 | ||||
AK165183.1 | - | 10 | 1539 | |||
AY101177.1 | - | 9 | 1446 | |||
BC062900.1 | - | 10 | 1584 | |||
BC141086.1 | - | 9 | 1380 | 9055698 |