Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028074.4 | NP_082350.3 | 15 | 2032 | ||
GenBank mRNA | AK007805.1 | - | 2 | 280 | ||
AK049311.1 | - | 15 | 2034 | |||
AK134182.1 | - | 14 | 1838 | |||
AK169816.1 | - | 15 | 1978 | |||
AK171730.1 | - | 15 | 1901 | |||
BC043036.1 | - | 14 | 1789 | 5698965 | ||
BC049382.1 | - | 3 | 444 |