Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028102.1 | NP_082378.1 | 14 | 1750 | ||
GenBank mRNA | AK079311.1 | - | 13 | 1731 | ||
AK173010.1 | - | 11 | 2058 | |||
BC046229.1 | - | 14 | 1670 | |||
BC118962.2 | - | 12 | 1743 | 40095026 |