Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028108.3 | NP_082384.1 | 3 | 481 | ||
GenBank mRNA | AK011160.1 | - | 3 | 407 | ||
AK135658.1 | - | 3 | 424 | |||
AK145345.1 | - | 3 | 409 | |||
AK156791.1 | - | 3 | 427 | |||
AK167141.1 | - | 4 | 601 | |||
AK168394.1 | - | 3 | 452 | |||
AK184762.1 | - | 2 | 177 | |||
AK195449.1 | - | 2 | 177 | |||
AK198408.1 | - | 3 | 250 | |||
AK219650.1 | - | 2 | 177 | |||
BC004086.1 | - | 3 | 393 | |||
BC055846.1 | - | 3 | 246 | 3600545 | ||
BC057117.1 | - | 3 | 440 | 6409354 |