Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028108.3 | NP_082384.1 | 2 | 358 | ||
GenBank mRNA | AK011160.1 | - | 2 | 284 | ||
AK135658.1 | - | 2 | 301 | |||
AK145345.1 | - | 2 | 286 | |||
AK156791.1 | - | 2 | 304 | |||
AK167141.1 | - | 3 | 478 | |||
AK168394.1 | - | 2 | 329 | |||
AK184762.1 | - | 1 | 54 | |||
AK195449.1 | - | 1 | 54 | |||
AK219650.1 | - | 1 | 54 | |||
BC004086.1 | - | 2 | 271 | |||
BC046283.1 | - | 2 | 278 | 6438976 | ||
BC055846.1 | - | 2 | 123 | 3600545 | ||
BC057117.1 | - | 2 | 317 | 6409354 | ||
BY062588.1 | - | 2 | 365 |