Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Seh1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039088.1 | NP_001034177.1 | 8 | 1074 | ||
NM_028112.2 | NP_082388.1 | 8 | 1074 | |||
GenBank mRNA | AK011330.1 | - | 8 | 976 | ||
AK088835.1 | - | 8 | 948 | |||
AK143727.1 | - | 8 | 1074 | |||
AK166573.1 | - | 8 | 939 | |||
AK169231.1 | - | 8 | 955 | |||
AK169388.1 | - | 8 | 954 | |||
AK170591.1 | - | 8 | 1027 | |||
BC027244.1 | - | 8 | 1026 | 3584697 |