Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028128.1 | NP_082404.1 | 8 | 776 | ||
GenBank mRNA | AK011489.1 | - | 8 | 787 | ||
AK150036.1 | - | 789 | ||||
AK170211.1 | - | 8 | 794 | |||
AK171372.1 | - | 776 | ||||
BC023674.1 | - | 7 | 714 | |||
BC089001.1 | - | 8 | 776 | 6415420 |