Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028135.2 | NP_082411.1 | 5 | 875 | ||
GenBank mRNA | AK011522.1 | - | 5 | 620 | ||
AK042650.1 | - | 5 | 624 | |||
AK047006.1 | - | 5 | 618 | |||
BC046234.1 | - | 875 | 6816957 | |||
BC059048.1 | - | 875 | 6816957 |