Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supt7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028150.2 | NP_082426.1 | 2 | 397 | ||
GenBank mRNA | AK012134.1 | - | 2 | 343 | ||
AK047063.1 | - | 2 | 367 | |||
AK139794.1 | - | 435 | ||||
AK144989.1 | - | 2 | 269 | |||
BC031447.1 | - | 2 | 320 | 4502314 |