Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152812.3 | NP_690025.2 | 4 | 654 | ||
GenBank mRNA | AK011203.1 | - | 5 | 585 | ||
AK144761.1 | - | 4 | 477 | |||
AK155761.1 | - | 4 | 505 | |||
AK166445.1 | - | 4 | 484 | |||
AK166546.1 | - | 4 | 481 | |||
AK169134.1 | - | 4 | 469 | |||
AK172229.1 | - | 4 | 459 | |||
AK172338.1 | - | 4 | 459 | |||
AK212698.1 | - | 1 | 94 | |||
BC031474.1 | - | 4 | 460 | 2599116 | ||
BC087552.1 | - | 4 | 451 | 6331952 |