Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152812.3 | NP_690025.2 | 5, 6 | 918 | ||
GenBank mRNA | AK011203.1 | - | 6, 7 | 849 | ||
AK144761.1 | - | 5, 6 | 741 | |||
AK155761.1 | - | 5, 6 | 769 | |||
AK166445.1 | - | 5, 6 | 748 | |||
AK169134.1 | - | 5, 6 | 733 | |||
AK172229.1 | - | 5, 6 | 723 | |||
AK172338.1 | - | 5, 6 | 723 | |||
BC031474.1 | - | 5, 6 | 724 | 2599116 | ||
BC087552.1 | - | 5, 6 | 715 | 6331952 |