Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007839.1 | - | 2 | 201 | ||
AK028304.1 | - | 9 | 998 | |||
AK088814.1 | - | 9 | 988 | |||
AK142658.1 | - | 9 | 988 | |||
AK145291.1 | - | 9 | 983 | |||
AK146207.1 | - | 9 | 987 | |||
AK150551.1 | - | 9 | 994 | |||
AK150777.1 | - | 9 | 952 | |||
AK151240.1 | - | 9 | 951 | |||
AK151409.1 | - | 9 | 994 | |||
AK151664.1 | - | 9 | 994 | |||
AK152338.1 | - | 9 | 994 | |||
AK152343.1 | - | 9 | 953 | |||
AK152991.1 | - | 9 | 953 | |||
AK153379.1 | - | 9 | 994 | |||
AK155905.1 | - | 9 | 988 | |||
AK159203.1 | - | 9 | 987 | |||
AK159350.1 | - | 9 | 987 | |||
AK159878.1 | - | 9 | 997 | |||
AK168708.1 | - | 9 | 930 | |||
AY029764.1 | - | 9 | 880 | |||
BC012401.1 | - | 9 | 977 | 3967323 | ||
CT010238.1 | - | 9 | 845 |