Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccpg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114328.2 | NP_001107800.1 | 8 | 1172 | ||
NM_001286544.1 | NP_001273473.1 | 8 | 1172 | |||
NM_001286545.1 | NP_001273474.1 | 8 | 1172 | |||
NM_001286546.1 | NP_001273475.1 | 8 | 1158 | |||
NM_028181.5 | NP_082457.3 | 8 | 1172 | |||
GenBank mRNA | AK032645.1 | - | 8 | 1044 | ||
AK076325.2 | - | 8 | 1099 | |||
AK128894.1 | - | 8 | 1098 | |||
AK134675.1 | - | 8 | 1069 | |||
BC006717.1 | - | 1 | 54 | |||
BC043049.1 | - | 8 | 1071 | 5704185 | ||
BC082602.1 | - | 8 | 1070 | 30663407 |