Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172073.1 | NP_001165544.1 | 7, 8 | 983 | ||
NM_028209.2 | NP_082485.1 | 7, 8 | 982 | |||
GenBank mRNA | AK005923.1 | - | 7, 8 | 983 | ||
AK088684.1 | - | 7, 8 | 944 | |||
AK133931.1 | - | 7, 8 | 947 | |||
BC025435.1 | - | 7, 8 | 931 | 5004463 | ||
BC125257.1 | - | 2, 3 | 320 |