Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197999.2 | NP_932116.2 | 6, 7 | 987 | ||
GenBank mRNA | AK146261.1 | - | 6, 7 | 931 | ||
BC027185.1 | - | 6, 7 | 965 | 4194872 |