Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2210408I21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081353.1 | NP_001074822.1 | 2 | 145 | ||
NM_001145676.1 | NP_001139148.1 | 2 | 145 | |||
GenBank mRNA | AK008859.1 | - | 2 | 131 | ||
AK050886.1 | - | 2 | 131 | |||
AK143619.1 | - | 2 | 83 | |||
AK165312.1 | - | 2 | 142 | |||
BB649442.1 | - | 2 | 145 | |||
BC089469.1 | - | 2 | 127 | |||
BC151066.1 | - | 2 | 127 | 9087979 | ||
BC157949.1 | - | 2 | 127 | 9088124 |