Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081180.1 | NP_001074649.1 | 9, 10 | 865 | ||
GenBank mRNA | AJ426449.2 | - | 9, 10 | 863 | ||
AK035690.1 | - | 9, 10 | 866 | |||
AK036804.1 | - | 9, 10 | 866 | |||
AK162690.1 | - | 9, 10 | 866 | |||
BC118011.1 | - | 9, 10 | 837 | 40097427 |