Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028238.7 | NP_082514.4 | 2 | 583 | ||
GenBank mRNA | AB232634.1 | - | 2 | 454 | ||
AK044783.1 | - | 2 | 546 | |||
AK054253.1 | - | 2 | 584 | |||
AK165682.1 | - | 2 | 586 | |||
AK196794.1 | - | 1 | 214 | |||
AK197116.1 | - | 1 | 214 | |||
AY062237.1 | - | 2 | 543 | |||
BC089578.1 | - | 2 | 543 | 6762755 |