Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114332.1 | NP_001107804.1 | 3 | 851 | ||
NM_028247.4 | NP_082523.1 | 3 | 851 | |||
GenBank mRNA | AK036743.1 | - | 3 | 852 | ||
AK050229.1 | - | 3 | 854 | |||
AK144135.1 | - | 3 | 854 | |||
AK151636.1 | - | 3 | 853 | |||
AK151704.1 | - | 3 | 856 | |||
AK157355.1 | - | 3 | 840 | |||
AK165240.1 | - | 3 | 852 | |||
AK165579.1 | - | 3 | 854 | |||
BC052877.1 | - | 3 | 332 | 30046388 |