Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114332.1 | NP_001107804.1 | 1 | 533 | ||
NM_028247.4 | NP_082523.1 | 1 | 533 | |||
GenBank mRNA | AK011813.1 | - | 1 | 537 | ||
AK036743.1 | - | 1 | 534 | |||
AK050229.1 | - | 1 | 536 | |||
AK144135.1 | - | 1 | 536 | |||
AK151290.1 | - | 1 | 534 | |||
AK151636.1 | - | 1 | 534 | |||
AK151704.1 | - | 1 | 538 | |||
AK157355.1 | - | 1 | 522 | |||
AK165240.1 | - | 1 | 534 | |||
AK165579.1 | - | 1 | 536 |