Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175639.1 | NP_783570.1 | 12 | 1535 | ||
GenBank mRNA | AK012043.1 | - | 12 | 1520 | ||
AK032907.1 | - | 12 | 1509 | |||
AK129031.1 | - | 12 | 1453 | |||
AK152321.1 | - | 12 | 1525 | |||
BC079913.1 | - | 11 | 1236 | |||
BC117724.2 | - | 13 | 1736 |