Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025392.1 | NP_001020563.1 | 7 | 2116 | ||
NM_001025393.1 | NP_001020564.1 | 7 | 2116 | |||
NM_153787.2 | NP_722482.1 | 7 | 2110 | |||
GenBank mRNA | AK079110.1 | - | 3 | 366 | ||
AK088741.1 | - | 7 | 2051 | |||
AK129071.1 | - | 4 | 1400 | |||
AK141834.1 | - | 7 | 2063 | |||
AK149724.1 | - | 7 | 2069 | |||
AK150017.1 | - | 7 | 2069 | |||
AK161608.1 | - | 1005 | ||||
AK161692.1 | - | 7 | 2114 | |||
BC029682.1 | - | 7 | 2047 | |||
BC034300.1 | - | 7 | 2042 | 4507876 | ||
BC086624.1 | - | 7 | 2098 |