Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110142.1 | NP_001103612.1 | 9 | 1381 | ||
NM_028288.5 | NP_082564.3 | 9 | 1322 | |||
GenBank mRNA | AB093259.1 | - | 6 | 643 | ||
AK012410.1 | - | 7 | 1085 | |||
AK032701.1 | - | 9 | 1323 | |||
AK160998.1 | - | 7 | 1092 | |||
AK170632.1 | - | 6 | 1470 | |||
AY330868.1 | - | 9 | 1323 | |||
BC004026.1 | - | 9 | 1370 | 3494708 |