Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110142.1 | NP_001103612.1 | 14, 15 | 1986 | ||
NM_028288.5 | NP_082564.3 | 14, 15 | 1927 | |||
GenBank mRNA | AB093259.1 | - | 10, 11 | 1544 | ||
AK012410.1 | - | 12, 13 | 1690 | |||
AK032701.1 | - | 14, 15 | 1928 | |||
AK160998.1 | - | 12, 13 | 1697 | |||
AY330868.1 | - | 14, 15 | 1928 | |||
BC004026.1 | - | 14, 15 | 1975 | 3494708 | ||
BC010347.1 | - | 1, 2 | 113 |