Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028315.2 | NP_082591.2 | 6, 7 | 1085 | ||
GenBank mRNA | AK032091.1 | - | 1, 2 | 175 | ||
AK038313.1 | - | 6, 7 | 955 | |||
AK129259.1 | - | 3, 4 | 1373 | |||
BC027357.1 | - | 6, 7 | 1055 | 4952415 |