Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164583.1 | NP_001158055.1 | 9 | 1213 | ||
NM_001164584.1 | NP_001158056.1 | 9 | 1158 | |||
NM_001164585.1 | NP_001158057.1 | 10 | 1223 | |||
NM_198412.2 | NP_940804.1 | 10 | 1132 | |||
GenBank mRNA | AK122293.1 | - | 9 | 1134 | ||
AK147570.1 | - | 10 | 1095 | |||
AK147637.1 | - | 10 | 1095 | |||
AK163657.1 | - | 9 | 1032 | |||
BC035262.1 | - | 4 | 367 | |||
BC060734.1 | - | 9 | 1213 | 6409942 | ||
BC064460.1 | - | 10 | 1101 | 6821822 |