Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028335.2 | NP_082611.1 | 6 | 1096 | ||
GenBank mRNA | AK012874.1 | - | 5 | 1047 | ||
BC049087.1 | - | 3 | 614 | |||
BC082571.1 | - | 6 | 1096 | 30615600 |