Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177323.3 | NP_796297.2 | 8 | 1184 | ||
GenBank mRNA | AK028707.1 | - | 7 | 1041 | ||
AK036805.1 | - | 8 | 1184 | |||
AK133789.1 | - | 8 | 1210 | |||
AK135334.1 | - | 8 | 1205 | |||
AK144144.1 | - | 8 | 1206 | |||
AK193380.1 | - | 1 | 104 | |||
BC065059.1 | - | 8 | 1184 | 6401664 | ||
BC145267.1 | - | 7 | 1000 | 9053821 | ||
BC150745.1 | - | 7 | 1000 | 9087656 |