Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028355.3 | NP_082631.1 | 2 | 458 | ||
GenBank mRNA | AK013409.1 | - | 2 | 195 | ||
AK144734.1 | - | 2 | 459 | |||
BC021337.1 | - | 2 | 213 | 5042118 | ||
DQ141695.1 | - | 2 | 153 |