Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pard3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081050.2 | NP_001074519.2 | 3 | 753 | ||
GenBank mRNA | AK008055.1 | - | 3 | 577 | ||
AK013352.1 | - | 3 | 754 | |||
AK141451.1 | - | 3 | 692 |