Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145123.4 | NP_660105.3 | 14 | 1908 | ||
GenBank mRNA | AJ421516.1 | - | 14 | 1758 | ||
AK013349.1 | - | 1 | 38 | |||
AK032328.1 | - | 14 | 1773 | |||
AK049801.1 | - | 14 | 1879 | |||
AK083221.1 | - | 14 | 1912 | |||
AK149020.1 | - | 14 | 1914 | |||
AK149069.1 | - | 14 | 1909 | |||
BC024472.1 | - | 11 | 1344 | 4976509 |