Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pot1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028370.1 | NP_082646.1 | 15 | 1871 | ||
GenBank mRNA | AK045286.1 | - | 15 | 1881 | ||
AK050727.1 | - | 15 | 1870 | |||
AK165040.1 | - | 15 | 1848 |