Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278415.1 | NP_001265344.1 | 3, 4 | 304 | ||
NM_028388.3 | NP_082664.1 | 2, 3 | 207 | |||
GenBank mRNA | AK013511.1 | - | 2, 3 | 158 | ||
AK078351.1 | - | 2, 3 | 151 | |||
AK146998.1 | - | 2, 3 | 141 | |||
AK150404.1 | - | 3, 4 | 241 | |||
AK151729.1 | - | 3, 4 | 241 | |||
AK159460.1 | - | 2, 3 | 124 | |||
AK166539.1 | - | 2, 3 | 168 | |||
AK169143.1 | - | 2, 3 | 178 | |||
BC030946.1 | - | 2, 3 | 136 | 5003524 | ||
BY246389.1 | - | 2, 3 | 208 | |||
CF106105.1 | - | 1, 2 | 49 |