Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top1mt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028404.2 | NP_082680.1 | 4 | 403 | ||
GenBank mRNA | AF362952.1 | - | 4 | 404 | ||
AK147119.1 | - | 4 | 406 | |||
BC138582.1 | - | 4 | 395 | 8861602 | ||
BC138583.1 | - | 4 | 395 | 8861603 | ||
BC145336.1 | - | 4 | 395 | 9053892 |