Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top1mt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028404.2 | NP_082680.1 | 8 | 1122 | ||
GenBank mRNA | AF362952.1 | - | 8 | 1123 | ||
AK013665.1 | - | 2 | 257 | |||
AK147119.1 | - | 8 | 1125 | |||
BC138582.1 | - | 8 | 1114 | 8861602 | ||
BC138583.1 | - | 8 | 1114 | 8861603 | ||
BC145336.1 | - | 8 | 1111 | 9053892 |