Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccser2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027045.1 | NP_081321.1 | 7 | 2391 | ||
NM_028407.3 | NP_082683.2 | 5 | 563 | |||
GenBank mRNA | AK005931.1 | - | 5 | 563 | ||
AK013689.1 | - | 7 | 2345 | |||
AK129295.1 | - | 6 | 1963 | |||
AK147383.1 | - | 7 | 2386 | |||
AK172153.1 | - | 7 | 2365 | |||
BC048641.1 | - | 5 | 562 | |||
BC060947.1 | - | 5 | 562 | |||
BC111465.1 | - | 6 | 772 | |||
BC139090.1 | - | 6 | 2179 | 8862112 |